missing translation for 'onlineSavingsMsg'
Learn More

BMP-3b/GDF-10 Antibody, Novus Biologicals™

Product Code. 18336078 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18336078 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18336078 Supplier Novus Biologicals Supplier No. H00002662B01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

BMP-3b/GDF-10 Polyclonal antibody specifically detects BMP-3b/GDF-10 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen BMP-3b/GDF-10
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS (pH 7.4)
Gene Accession No. NP_004953
Gene Alias BIP, BMP3B, BMP-3B, bone morphogenetic protein 3B, Bone-inducing protein, GDF-10, growth differentiation factor 10, Growth/differentiation factor 10
Host Species Mouse
Immunogen GDF10 (NP_004953, 1 a.a. - 478 a.a.) full-length human protein. MAHVPARTSPGPGPQLLLLLLPLFLLLLRDVAGSHRAPAWSALPAAADGLQGDRDLQRHPGDAAATLGPSAQDMVAVHMHRLYEKYSRQGARPGGGNTVRSFRARLEVVDQKAVYFFNLTSMQDSEMILTATFHFYSEPPRWPRALEVLCKPRAKNASGRPLPLGPPTRQHLLFRSLSQNTATQGLLRGAMALAPPPRGLWQAKDISPIVKAARRDGELLLSAQLDSEERDPGVPRPSPYAPYILVYANDLAISEPNSVAVTLQRYDPFPAGDPEPRAAPNNSADPRVRRAAQATGPLQDNELPGLDERPPRAHAQHFHKHQLWPSPFRALKPRPGRKDRRKKGQEVFMAASQVLDFDEKTMQKARRKQWDEPRVCSRRYLKVDFADIGWNEWIISPKSFDAYYCAGACEFPMPKIVRPSNHATIQSIVRAVGIIPGIPEPCCVPDKMNSLGVLFLDENRNVVLKVYPNMSVDTCACR
Purification Method Protein A purified
Quantity 0.05 mg
Regulatory Status RUO
Research Discipline Cytokine Research
Primary or Secondary Primary
Gene ID (Entrez) 2662
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.