Learn More
PeproTech BMP-2 Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™

Rabbit Polyclonal Antibody
Brand: PeproTech 500-P195BT-25UG
Description
AA Sequence of recombinant protein: MQAKHKQRKR LKSSCKRHPL YVDFSDVGWN DWIVAPPGYH AFYCHGECPF PLADHLNSTN HAIVQTLVNS VNSKIPKACC VPTELSAISM LYLDENEKVV LKNYQDMVVE GCGCR. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human BMP-2. Anti-Human BMP-2-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect hBMP-2 by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Human BMP-2 (500-P195) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant hBMP-2. Western Blot: To detect Human BMP-2 by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 mg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant Human BMP-2 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P195BT-1MG will be provided as 2 x 500 μg
Bone Morphogenic Proteins (BMP) are members of the TGF-beta superfamily that affect bone and cartilage formation (Hogan 1996, Reddi 1998 and Francis-West et al. 1999). Mature BMPs are 30-38 kDa proteins that assume a TGF-beta -like cysteine knot configuration. Lovostatin increases bone formation by turning on the bmp-2 gene (Mundy et al. 1999). BMPs stimulate the production of specific bone matrix proteins and alter stromal cell and osteoclast proliferation (Macias et al. 1999, Lecanda et al. 1997). BMPs may also be an important factor for development of the viscera, with roles in cell proliferation, apoptosis, differentiation, and morphogenesis (Hogan 1996, Dale and Wardle 1999). BMPs appear to be responsible for normal dorsal/ventral patterning. Like TGF-beta, BMPs bind to a type II receptor, which then recruits the transducing type I receptor unit, activating the Smad protein signaling pathway (Massague 1994, Derynck 1997, Attisano 1993).
Specifications
| BMP-2 | |
| Polyclonal | |
| Biotin | |
| BMP2 | |
| 2610024H22Rik; AI467020; AL117858; AW546137; BB189135; BDA2; BMP; BMP2; BMP-2; BMP2A; BMP-2A; BMPRII; BMPR-II; Bone morphogenetic; Bone morphogenetic protein; Bone morphogenetic protein 2; bone morphogenetic protein 2 precursor (BMP-2) (BMP-2A); bone morphogenetic protein 2A; H-BMP-2; morphogen | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 12156, 29373, 650 | |
| -20°C | |
| Lyophilized |
| ELISA, Western Blot | |
| 0.1-1.0 mg/mL | |
| PBS with no preservative | |
| P12643, P21274, P49001 | |
| BMP2 | |
| E.coli-derived Recombinant Human/Murine/Rat BMP-2. | |
| 25 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.