missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
BMP-2/BMP-4 Polyclonal antibody specifically detects BMP-2/BMP-4 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunoprecipitation, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | BMP-2/BMP-4 |
| Applications | Western Blot, ELISA, Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500, ELISA 1:2000, Immunoprecipitation 1:200, Immunohistochemistry-Paraffin 1:50-1:150 |
| Formulation | Contains Tris, HCl/glycine buffer (pH 7.4-7.8), 30% glycerol, 0.5% BSA, along with cryo-protective agents, and HEPES (exact storage and stabilization buffer information is proprietary) with 0.02% Sodium Azide |
| Gene Alias | BDA2, BMP-2, BMP2A, BMP-2A, BMP2ABone morphogenetic protein 2A, bone morphogenetic protein 2 |
| Host Species | Rabbit |
| Immunogen | Synthetic peptide taken within amino acid region 184-197 on rat BMP4 protein, peptide was covalently modified. Sequence: VMKPPAPGHLIC |
| Purification Method | Affinity-purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?