missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BMP-2 Antibody (9E10G12) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£196.00 - £471.00
Specifications
| Antigen | BMP-2 |
|---|---|
| Clone | 9E10G12 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200-1:1000 |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18199819
|
Novus Biologicals
NBP2-52422 |
0.1 mg |
£471.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18652638
|
Novus Biologicals
NBP2-52422-0.025mg |
0.025 mg |
£196.00
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
BMP-2 Monoclonal specifically detects BMP-2 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| BMP-2 | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Biologically Active Proteins, Cytokine Research, Embryonic Stem Cell Markers, Immunology, Neuronal Cell Markers, Neuronal Stem Cell Markers, Stem Cell Markers, Stem Cell Signaling Pathway | |
| PBS with 0.05% Sodium Azide | |
| 650 | |
| Purified recombinant fragment of human BMP2 (AA: 283-396) expressed in E. Coli. | |
| Primary | |
| 44.7 kDa |
| 9E10G12 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200-1:1000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| BMP-2, BMP-2A, BMP2ABone morphogenetic protein 2A, bone morphogenetic protein 2 | |
| BMP2 | |
| IgG1 | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title