missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BMP-15 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA534401
This item is not returnable.
View return policy
Description
A suggested positive control is NIH-3T3 cell lysate. PA5-34401 can be used with blocking peptide PEP-1443.
BMP15, or GDF9B is a member of the bone morphogenetic protein family which is part of the transforming growth factor-beta superfamily that are involved in embryonic development and adult tissue homeostasis. BMP15 is expressed exclusively in the oocyte. It is an oocyte-specific growth/differentiation factor that may be involved in oocyte maturation and follicular development. Defects in BMP15 are the cause of ovarian dysgenesis type 2. BMP15 could be used as an oogenesis marker to track human amniotic fluid stem cells differentiation into the oocyte-like cells.
Specifications
| BMP-15 | |
| Polyclonal | |
| Unconjugated | |
| BMP15 | |
| AU015375; AU018861; AU021453; BMP; BMP15; Bmp-15; Bone morphogenetic protein; Bone morphogenetic protein 15; C86824; C87336; GDF9B; GDF-9B; growth differentiation factor-9B; growth/differentiation factor 9B; ODG2; POF4 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 12155, 9210 | |
| Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| O95972, Q9Z0L4 | |
| BMP15 | |
| A 16 amino acid synthetic peptide near the amino terminus of human BMP15. | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction