missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BMP-15/GDF-9B Mouse anti-Human, Clone: BMP15/4263, Novus Biologicals™
Mouse Monoclonal Antibody
£196.00
Specifications
| Antigen | BMP-15/GDF-9B |
|---|---|
| Clone | BMP15/4263 |
| Concentration | 0.2 mg/ml |
| Dilution | Western Blot 1-2 ug/ml, ELISA |
| Applications | Western Blot, ELISA |
Description
BMP-15/GDF-9B Monoclonal specifically detects BMP-15/GDF-9B in Human samples. It is validated for Western Blot, ELISA.Specifications
| BMP-15/GDF-9B | |
| 0.2 mg/ml | |
| Western Blot, ELISA | |
| Unconjugated | |
| Mouse | |
| Human | |
| BMP-15, bone morphogenetic protein 15, GDF-9B, Growth/differentiation factor 9B, ODG2, POF4 | |
| Recognizes sequence SAEVTASSSKHSGPENNQC on the C-terminus of human BMP-15/GDF-9B | |
| Primary | |
| Store at 4°C. |
| BMP15/4263 | |
| Western Blot 1-2 ug/ml, ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10 mM PBS with 0.05% BSA | |
| 9210 | |
| IgG1 | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title