missing translation for 'onlineSavingsMsg'
Learn More

BMP-15/GDF-9B Mouse anti-Human, Clone: BMP15/4263, Novus Biologicals™

Mouse Monoclonal Antibody

£377.00

Specifications

Antigen BMP-15/GDF-9B
Clone BMP15/4263
Concentration 0.2 mg/mL
Dilution Western Blot 1 to 2 μg/mL, ELISA
Applications Western Blot, ELISA
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18699038
View Documents
Novus Biologicals
NBP3-07552-100ug
100 μg
£377.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

BMP-15/GDF-9B Monoclonal specifically detects BMP-15/GDF-9B in Human samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

BMP-15/GDF-9B
0.2 mg/mL
Western Blot, ELISA
Unconjugated
Mouse
Human
BMP-15, bone morphogenetic protein 15, GDF-9B, Growth/differentiation factor 9B, ODG2, POF4
Recognizes sequence SAEVTASSSKHSGPENNQC on the C-terminus of human BMP-15/GDF-9B
Primary
Store at 4°C.
BMP15/4263
Western Blot 1 to 2 μg/mL, ELISA
Monoclonal
Purified
RUO
10 mM PBS with 0.05% BSA
9210
IgG1
Protein A or G purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.