missing translation for 'onlineSavingsMsg'
Learn More

BMP-15/GDF-9B Antibody (317809) [DyLight 755], Novus Biologicals™

Product Code. 30737475 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30737475 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30737475 Supplier Novus Biologicals Supplier No. FAB2925Z

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

BMP-15/GDF-9B Monoclonal antibody specifically detects BMP-15/GDF-9B in Human samples. It is validated for Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen BMP-15/GDF-9B
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 317809
Conjugate DyLight 755
Formulation 50mM Sodium Borate
Gene Alias BMP-15, bone morphogenetic protein 15, GDF-9B, Growth/differentiation factor 9B, ODG2, POF4
Host Species Rat
Immunogen E. coli-derived recombinant human BMP-15/GDF-9B, Gln268-Arg392, Accession # O95972
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Cardiovascular Biology, Cell Biology, Cytokines and Growth Factors, Immunology, Signal Transduction, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 9210
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.