missing translation for 'onlineSavingsMsg'
Learn More

BMP-10 Antibody (462732) [Janelia Fluor™ 646], Novus Biologicals™

Product Code. 30738248 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30738248 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30738248 Supplier Novus Biologicals Supplier No. FAB2926J

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

BMP-10 Monoclonal antibody specifically detects BMP-10 in Human,Mouse samples. It is validated for Western Blot,Neutralization
TRUSTED_SUSTAINABILITY

Specifications

Antigen BMP-10
Applications Western Blot, Neutralization
Classification Monoclonal
Clone 462732
Conjugate Janelia Fluor 646
Formulation 50mM Sodium Borate
Gene Alias BMP-10, bone morphogenetic protein 10, MGC126783
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human BMP-10, Asn317-Arg424, Accession # O95393
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 27302
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.