missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BMP-1/PCP Antibody (OTI3E9), DyLight 550, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70255R
This item is not returnable.
View return policy
Description
BMP-1/PCP Monoclonal specifically detects BMP-1/PCP in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| BMP-1/PCP | |
| Monoclonal | |
| DyLight 550 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| BMP1 | |
| Recombinant protein fragment corresponding to amino acids 225-617 of human BMP-1/PCP (NP_001190) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| OTI3E9 | |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| BMP-1, bone morphogenetic protein 1, EC 3.4.24, EC 3.4.24.19, FLJ44432, Mammalian tolloid protein, mTld, PCOLC, PCP, PCP2, procollagen C-endopeptidase, Procollagen C-proteinase, TLD | |
| Mouse | |
| Affinity Purified | |
| Cytokine Research | |
| 649.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction