missing translation for 'onlineSavingsMsg'
Learn More

BML-284, MedChemExpress

Product Code. 16440468
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16440468 5 mg 5mg
16473927 50 mg 50mg
16417786 100 mg 100mg
16408008 10 mg 10mg
16401306 200 mg 200mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16440468 Supplier Medchem Express Supplier No. HY19987

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress BML-284 is a potent and cell-permeable Wnt signaling activator. BML-284 induces TCF-dependent transcriptional activity with an EC50 of 700 nM.

BML-284 is a potent and cell-permeable Wnt signaling activator. BML-284 induces TCF-dependent transcriptional activity with an EC50 of 700 nM.

Specifications

Chemical Name or Material BML-284
CAS 853220-52-7
Color White
Purity Grade Notes Research
Molecular Formula C19H18N4O3
Quantity 5 mg
Solubility Information DMSO : ≥ 100 mg/mL (285.41 mM)
SMILES NC1=NC(C2=CC=CC(OC)=C2)=CC(NCC3=CC=C(OCO4)C4=C3)=N1
Molecular Weight (g/mol) 350.37
Formula Weight 350.37
Percent Purity 97.48%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.