missing translation for 'onlineSavingsMsg'
Learn More

HLY78, MedChemExpress

Product Code. 16417787
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
This item is not returnable. View return policy

Product Code. 16417787

Brand: Medchem Express HY122816

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress HLY78 is an activator of the Wnt/β-catenin signaling pathway, which targets the DIX domain of Axin and potentiates the Axin-LRP6 association to promote Wnt signaling transduction.

HLY78 is an activator of the Wnt/β-catenin signaling pathway, which targets the DIX domain of Axin and potentiates the Axin-LRP6 association to promote Wnt signaling transduction.

Specifications

Chemical Name or Material HLY78
CAS 854847-61-3
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C17H17NO2
Quantity 5 mg
Solubility Information DMSO : 26 mg/mL (97.26 mM; Need ultrasonic)
SMILES CCC1=CC=CC2=C1N(CC3=CC4=C(C=C32)OCO4)C
Molecular Weight (g/mol) 267.32
Formula Weight 267.32
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.