missing translation for 'onlineSavingsMsg'
Learn More

BML-190, MedChemExpress

Product Code. 16412607
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
This item is not returnable. View return policy

Product Code. 16412607

Brand: Medchem Express HY15420

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress BML-190(IMMA) is a potent and selective CB2 receptor ligand (Ki values are 435 nM and > 2 μM for CB2 and CB1 respectively).

BML-190(IMMA) is a potent and selective CB2 receptor ligand (Ki values are 435 nM and > 2 μM for CB2 and CB1 respectively).

Specifications

Chemical Name or Material BML-190
CAS 2854-32-2
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C23H23ClN2O4
Quantity 50 mg
Synonym Indomethacin morpholinylamide IMMA
Solubility Information DMSO : 50 mg/mL (117.13 mM; Need ultrasonic)
SMILES CC(N1C(C2=CC=C(Cl)C=C2)=O)=C(CC(N3CCOCC3)=O)C4=C1C=CC(OC)=C4
Molecular Weight (g/mol) 426.89
Formula Weight 426.89
Percent Purity 99.54%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.