missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BMI-1 Recombinant Rabbit Monoclonal Antibody (23GB685), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA554178
This item is not returnable.
View return policy
Description
Component of the Polycomb group (PcG) multiprotein PRC1 complex, a complex required to maintain the transcriptionally repressive state of many genes, including Hox genes, throughout development. PcG PRC1 complex acts via chromatin remodeling and modification of histones; it mediates monoubiquitination of histone H2A 'Lys-119', rendering chromatin heritably changed in its expressibility. In the PRC1 complex, it is required to stimulate the E3 ubiquitin-protein ligase activity of RNF2/RING2.
Specifications
| BMI-1 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| BMI1 | |
| AW546694; B lymphoma Mo-MLV insertion region 1; B lymphoma Mo-MLV insertion region 1 homolog; Bmi1; Bmi-1; BMI1 polycomb ring finger oncogene; BMI1 polycomb ring finger proto-oncogene; BMI1 proto-oncogene, polycomb ring finger; FLVI2/BMI1; flvi-2/bmi-1; murine leukemia viral (bmi-1) oncogene homolog; Pcgf4; Pcgf4 antibody; Polycomb complex protein BMI-1; polycomb group protein Bmi1; polycomb group ring finger 4; polycomb group RING finger protein 4; RING finger protein 51; RNF51; RP11-573G6.1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 307151, 648 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB685 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P35226 | |
| BMI1 | |
| A synthetic peptide of human BMI1 (302-326AA). | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction