Learn More
Invitrogen™ BMI-1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA589253
Description
Immunogen sequence: EDKRIITDDE IISLSIEFFD QNRLDRKVNK DKEKSKEEVN DKRYLRCPAA MTVMHLRKFL RSKMDIPNTF QIDVMYEEEP LKDYYTLMDI AYIYTWRRNG PLPLKYRVRP TCKRMKISHQ RDGLTNAGEL ESDSGSDKAN SPAGGIPSTS SCLPSPSTPV QSPHPQFPHI SSTMNGTSNS PSGNHQSSFA NRPRKSSVNG SSATSSG; Positive Samples: U-937; Cellular Location: Cytoplasm, Nucleus.
Component of the Polycomb group (PcG) multiprotein PRC1 complex, a complex required to maintain the transcriptionally repressive state of many genes, including Hox genes, throughout development. PcG PRC1 complex acts via chromatin remodeling and modification of histones; it mediates monoubiquitination of histone H2A 'Lys-119', rendering chromatin heritably changed in its expressibility. In the PRC1 complex, it is required to stimulate the E3 ubiquitin-protein ligase activity of RNF2/RING2.
Specifications
| BMI-1 | |
| Polyclonal | |
| Unconjugated | |
| BMI1 | |
| AW546694; B lymphoma Mo-MLV insertion region 1; B lymphoma Mo-MLV insertion region 1 homolog; Bmi1; Bmi-1; BMI1 polycomb ring finger oncogene; BMI1 polycomb ring finger proto-oncogene; BMI1 proto-oncogene, polycomb ring finger; FLVI2/BMI1; flvi-2/bmi-1; murine leukemia viral (bmi-1) oncogene homolog; Pcgf4; Pcgf4 antibody; Polycomb complex protein BMI-1; polycomb group protein Bmi1; polycomb group ring finger 4; polycomb group RING finger protein 4; RING finger protein 51; RNF51; RP11-573G6.1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 648 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P35226 | |
| BMI1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 120-326 of human Bmi1 (NP_0051714). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.