missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BMI-1 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB110-40823CL1
This item is not returnable.
View return policy
Description
BMI-1 Polyclonal antibody specifically detects BMI-1 in Human,Mouse samples. It is validated for Immunohistochemistry,Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| BMI-1 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Immunohistochemistry, Western Blot, Immunocytochemistry/Immunofluorescence | |
| CoraFluor 1 | |
| B lymphoma Mo-MLV insertion region 1 homolog, B lymphoma Mo-MLV insertion region 1 homolog (mouse), BMI1 polycomb ring finger oncogene, FLVI2/BMI1, MGC12685, murine leukemia viral (bmi-1) oncogene homolog, PCGF4, polycomb complex protein BMI-1, polycomb group protein Bmi1, polycomb group ring finger 4, RING finger protein 51, RNF51Polycomb group RING finger protein 4 | |
| A synthetic peptide made to an internal region (within residues 1-100) of human Bmi1. [Swiss-Prot# P35226] | |
| 0.1 mL | |
| Breast Cancer, Cancer, Cancer Stem Cells, Cell Biology, Cell Cycle and Replication, Chromatin Research, DNA Repair, DNA replication Transcription Translation and Splicing, Epigenetics, Hematopoietic Stem Cell Markers, Stem Cell Markers, Transcription Factors and Regulators | |
| 648 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction