missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bmf Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76658
This item is not returnable.
View return policy
Description
Bmf Polyclonal specifically detects Bmf in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| Bmf | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| Bcl2 modifying factor, bcl-2-modifying factor, FLJ00065 | |
| Rabbit | |
| 22 kDa | |
| 0.1 mg | |
| Apoptosis, Cancer, Tumor Suppressors | |
| 90427 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| 1 mg/mL | |
| Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunofluorescence 5 ug/ml, Immunocytochemistry 10 ug/ml | |
| NP_277038 | |
| BMF | |
| Antibody was raised with a synthetic peptide corresponding to 15 amino acids near the amino terminus of human Bmf. The immunogen is located within the first 50 amino acids of Bmf. Amino Acid Squence: EPSQCVEELEDDV | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction