missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BMAL1 Recombinant Rabbit Monoclonal Antibody (HL2455), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen MA550536
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a basic helix-loop-helix protein that forms a heterodimer with CLOCK. This complex binds an E-box upstream of the PER1 gene, activating this gene and possibly other circadian rhythym-associated genes. Three transcript variants encoding two different isoforms have been found for this gene.
Specifications
| BMAL1 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| O00327 | |
| Arntl | |
| Recombinant fragment of human BMAL1. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Western Blot | |
| HL2455 | |
| Unconjugated | |
| Arntl | |
| Arnt3, ARNTL, ARNT-like protein 1, brain and muscle, aryl hydrocarbon receptor nuclear translocator like, aryl hydrocarbon receptor nuclear translocator-like, aryl hydrocarbon receptor nuclear translocator-like protein 1, basic-helix-loop-helix-PAS orphan MOP3, basic-helix-loop-helix-PAS protein MOP3, bHLHe5, bHLH-PAS protein JAP3, BMAL1, Bmal1 protein, BMAL1b, bmal1b', BMAL1c, Brain and muscle ARNT-like 1, brain and muscle Arnt-like protein 1, CG8727 PA, class E basic helix-loop-helix protein 5, cycle, JAP3, LOW QUALITY PROTEIN: aryl hydrocarbon receptor nuclear translocator-like protein 1, member of PAS protein 3, member of PAS superfamily 3, MGC47515, MOP3, PAS domain-containing protein 3, PASD3, testis tissue sperm-binding protein Li 50e, tic | |
| Rabbit | |
| Protein A | |
| RUO | |
| 406 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction