Learn More
Invitrogen™ BMAL1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA1523
Description
PA1-523 detects BMAL1/aryl hydrocarbon nuclear translocator 3 (ARNT3) from hamster and mouse tissues as well as recombinant human BMAL1. PA1-523 has been sucessfully used in Western blot procedures. By Western blot, this antibody detects a 110 kDa protein which corresponds to the product of a hamster GST-BMAL1 fusion construct overexpressed in E. coli. This antibody detects a non-specific band in U87-MG cell lysates at ~105kDa and in NIH-3T3 cell lysates at ~135kDa. PA1-523 immunizing peptide corresponds to amino acid residues 582-594 from mouse BMAL1. This sequence is completely conserved between mouse, rat, guinea pig, and human BMAL1. PA1-523 immunizing peptide (Cat. # PEP-075) is available for use in neutralization and control experiments.
The protein encoded by this gene is a basic helix-loop-helix protein that forms a heterodimer with CLOCK. This complex binds an E-box upstream of the PER1 gene, activating this gene and possibly other circadian rhythym-associated genes. Three transcript variants encoding two different isoforms have been found for this gene.
Specifications
| BMAL1 | |
| Polyclonal | |
| Unconjugated | |
| Arntl | |
| Arnt3; ARNTL; ARNT-like protein 1, brain and muscle; aryl hydrocarbon receptor nuclear translocator like; aryl hydrocarbon receptor nuclear translocator-like; aryl hydrocarbon receptor nuclear translocator-like protein 1; basic-helix-loop-helix-PAS orphan MOP3; basic-helix-loop-helix-PAS protein MOP3; bHLHe5; bHLH-PAS protein JAP3; BMAL1; Bmal1 protein; BMAL1b; bmal1b'; BMAL1c; Brain and muscle ARNT-like 1; brain and muscle Arnt-like protein 1; CG8727 PA; class E basic helix-loop-helix protein 5; cycle; JAP3; LOW QUALITY PROTEIN: aryl hydrocarbon receptor nuclear translocator-like protein 1; member of PAS protein 3; member of PAS superfamily 3; MGC47515; MOP3; PAS domain-containing protein 3; PASD3; testis tissue sperm-binding protein Li 50e; tic | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 100755141, 11865, 29657, 406 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 1mg/mL BSA and 0.05% sodium azide | |
| O00327, Q9EPW1, Q9WTL8 | |
| Arntl | |
| Synthetic Peptide: C D(582) M I D N D Q G S S S P S(594). | |
| 200 μL | |
| Primary | |
| Human, Mouse, Rat, Hamster | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.