missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BM212, MedChemExpress
Description
BM212 is a potent Mycobacterial membrane protein Large 3 (MmpL3) inhibitor. BM212 has strong bactericidal activity against both M. tuberculosis and some nontuberculosis mycobacteria. BM212 exhibits antimycobacterial activity against M. tuberculosis H37Rv with an MIC of 5 μM.
Specifications
Specifications
| Chemical Name or Material | BM212 |
| CAS | 146204-42-4 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C23H25Cl2N3 |
| Quantity | 5 mg |
| Solubility Information | Ethanol : 5.56 mg/mL (13.42 mM; Need ultrasonic) ∣DMSO : 1 mg/mL (2.41 mM; Need ultrasonic) |
| SMILES | CN1CCN(CC2=C(C)N(C3=CC=C(Cl)C=C3)C(C4=CC=C(Cl)C=C4)=C2)CC1 |
| Molecular Weight (g/mol) | 414.37 |
| Formula Weight | 414.37 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?