missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BLZF1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA582897
This item is not returnable.
View return policy
Description
Immunogen sequence: KSLGHHKGEF LGQSEGVIEP NKELSEVKNV LEKLKNSERR LLQDKEGLSN QLRVQTEVNR ELKKLLVASV GDDLQYHFER LARE Highest antigen sequence indentity to the following orthologs: Mouse - 85%, Rat - 87%.
Required for normal Golgi structure and for protein transport from the endoplasmic reticulum (ER) through the Golgi apparatus to the cell surface.
Specifications
| BLZF1 | |
| Polyclonal | |
| Unconjugated | |
| BLZF1 | |
| 1700030G05Rik; Basic leucine zipper nuclear factor 1; basic leucine zipper nuclear factor 1 (JEM-1); Blzf1; blzf1 protein; Blzf1l; cytoplasmic protein; fe06a04; Golgin45; Golgin-45; Golgin-45; golgin-45; JEM1; JEM-1; JEM-1s; JEM-1short protein; LOC504820 protein; MGC22497; p45 basic leucine-zipper nuclear factor; wu:fe06a04; zgc:66280; zgc:85871 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 8548 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9H2G9 | |
| BLZF1 | |
| Recombinant protein corresponding to Human BLZF1. Recombinant protein control fragment (Product #RP-94118). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction