missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BLZF1 Monoclonal Antibody (OTI8E8)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA527126
This item is not returnable.
View return policy
Description
BLZF1 Monoclonal Antibody for Western Blot
Required for normal Golgi structure and for protein transport from the endoplasmic reticulum (ER) through the Golgi apparatus to the cell surface.
Specifications
| BLZF1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9H2G9 | |
| BLZF1 | |
| Human recombinant protein fragment corresponding to amino acids 1-246 of BLZF1 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI8E8 | |
| Unconjugated | |
| BLZF1 | |
| 1700030G05Rik; Basic leucine zipper nuclear factor 1; basic leucine zipper nuclear factor 1 (JEM-1); Blzf1; blzf1 protein; Blzf1l; cytoplasmic protein; fe06a04; Golgin45; Golgin-45; Golgin-45; golgin-45; JEM1; JEM-1; JEM-1s; JEM-1short protein; LOC504820 protein; MGC22497; p45 basic leucine-zipper nuclear factor; wu:fe06a04; zgc:66280; zgc:85871 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 8548 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction