missing translation for 'onlineSavingsMsg'
Learn More

BLZF1 Antibody (OTI8E8), Novus Biologicals Biologicals™

Mouse Monoclonal Antibody

£195.00 - £417.00

Specifications

Antigen BLZF1
Clone OTI8E8
Dilution Western Blot 1:500-2000
Applications Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30306509
View Documents
Novus Biologicals
NBP3-44936-100ul
100 μL
£417.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30306348
View Documents
Novus Biologicals
NBP3-44936-30ul
30 μL
£195.00
30µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

BLZF1 Monoclonal antibody specifically detects BLZF1 in Human samples. It is validated for Western Blot
Specifications

Specifications

BLZF1
Western Blot 1:500-2000
Monoclonal
Purified
RUO
PBS (pH 7.3) 1% BSA, 50% glycerol
8548
IgG1
>95%
Antibody
OTI8E8
Western Blot
Unconjugated
Mouse
Human
basic leucine zipper nuclear factor 1JEM-1s, cytoplasmic protein, GOLGIN-45, JEM1, JEM-1MGC22497, JEM-1short protein, p45 basic leucine-zipper nuclear factor
Human recombinant protein fragment corresponding to amino acids 1-246 of human BLZF1(NP_003657) produced in E.coli.
Primary
Store at -20 degrees C. Avoid freeze/thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.