missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BLZF1 Antibody (OTI3E2), Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-44725
This item is not returnable.
View return policy
Description
BLZF1 Monoclonal antibody specifically detects BLZF1 in Human samples. It is validated for Western Blot
Specifications
| BLZF1 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized powder ( 1X PBS, pH 7.3,) | |
| Mouse | |
| >95% | |
| RUO | |
| 8548 | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI3E2 | |
| Western Blot 1:2000 | |
| basic leucine zipper nuclear factor 1JEM-1s, cytoplasmic protein, GOLGIN-45, JEM1, JEM-1MGC22497, JEM-1short protein, p45 basic leucine-zipper nuclear factor | |
| Human recombinant protein fragment corresponding to amino acids 1-246 of human BLZF1(NP_003657) produced in E.coli. | |
| 100 μg | |
| Primary | |
| For reconstitution, we recommend adding 100uL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiment, we strongly recommend performing another round of desalting process. | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction