missing translation for 'onlineSavingsMsg'
Learn More

BLZF1 Antibody, Novus Biologicals™

Product Code. 18352619 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18352619 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18352619 Supplier Novus Biologicals Supplier No. H00008548B01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

BLZF1 Polyclonal antibody specifically detects BLZF1 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen BLZF1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS (pH 7.4)
Gene Accession No. NP_003657.1
Gene Alias basic leucine zipper nuclear factor 1JEM-1s, cytoplasmic protein, GOLGIN-45, JEM1, JEM-1MGC22497, JEM-1short protein, p45 basic leucine-zipper nuclear factor
Host Species Mouse
Immunogen BLZF1 (NP_003657.1, 1 a.a. - 400 a.a.) full-length human protein. MTTKNLETKVTVTSSPIRGAGDGMETEEPPKSVEVTSGVQSRKHHSLQSPWKKAVPSESPGVLQLGKMLTEKAMEVKAVRILVPKAAITHDIPNKNTKVKSLGHHKGEFLGQSEGVIEPNKELSEVKNVLEKLKNSERRLLQDKEGLSNQLRVQTEVNRELKKLLVASVGDDLQYHFERLAREKNQLILENEALGRNTAQLSEQLERMSIQCDVWRSKFLASRVMADELTNSRAALQRQNRDAHGAIQDLLSEREQFRQEMIATQKLLEELLVSLQWGREQTYSPSVQPHSTAELALTNHKLAKAVNSHLLGNVGINNQKKIPSTVEFCSTPAEKMAETVLRILDPVTCKESSPDNPFFESSPTTLLATKKNIGRFHPYTRYENITFNCCNHCRGELIAL
Purification Method Protein G purified
Quantity 0.05 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8548
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.