Learn More
Melford Bluo-Gal
Description
Chromogenic substrate for B-D-Galactosidase.
Chromogenic substrate for B-D-Galactosidase. Produces a deeper blue water insoluble color than X-GAL.
Specifications
Specifications
| Quantity | 100 mg |