missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Blunt Fill Needles
Description
- Non coring bevel design to pierce IV bag septum rubber vial caps
- Eliminates risk of rubber particles being cut from multi-dose vial stoppers
Blunt Fill Needle:
- Allows rapid filling, even with viscous solutions
- Color coded for easy identification
Application:
- Contributes to safety of healthcare professionals
- Safely bridge medication preparation and administration
Certifications
EN ISO 7886-1
Specifications
Specifications
| Certifications/Compliance | CE marked and CE certified with AEMPS (0318) |
| Color | Red |
| Needle Gauge | 18 G |
| Length (Metric) | 40 mm |
| Product Type | Blunt Fill Needles |
| Quantity | 100/Box |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?