missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Shermond™ Blue Polythene Gauntlet Gloves
Click to view available options
:
Size: Universal
Description
- Fully disposable arm-length gauntlets
- Food safe polythene
- Ideal for general purpose use including food handling and also veterinary applications
- Ambidextrous fit
- Powder free and non-sterile
- Supplied in 50 packs of 50 gloves
- Not suitable for clinical applications
Specifications
Specifications
| Color | Blue |
| Exterior Finish | No Texture |
| Hand | Ambidextrous |
| Latex Free | Yes |
| Material | Polyethylene |
| For Use With (Application) | General Purpose use Including Food Hygiene |
| Glove Cuff Style | Gauntlet |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction