missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ Blue Disposable Visitor Lab Coats, Series 4400
Use for hygiene applications. The 3M Disposable Visitor Lab Coats series is made from lightweight polypropylene.
Brand: 3M™ 7000089701
Code : 94
Additional Details : Weight : 1.00000kg
Description
- Ideal for hygiene applications
- 100% soft, breathable, and lightweight polypropylene
- Zip fastener
- Internal pocket
Specifications
Lab Coat | |
Polypropylene | |
Zipper | |
Single-use | |
36 to 38 in. | |
CE Category 1 | |
Internal pocket on the wearers left hand side (measuring 13 H x 12cm W) |
Large | |
Blue | |
Elastic | |
Serged | |
91 to 97 cm | |
Non-hazardous liquids and dust | |
Individual packaging |