missing translation for 'onlineSavingsMsg'
Learn More

Syngene™ Blue Light Converter Screen
GREENER_CHOICE

Product Code. 15819892 Shop All Syngene Products
Change view
Click to view available options
For Use With (Equipment):
Gbox Mini, Nugenius, Ingenius
Unit Size:
Each
Product Code. For Use With (Equipment) unitSize
15819892 Gbox Mini, Nugenius, Ingenius Each
1 options
This item is not returnable. View return policy

Product Code. 15819892

Brand: Syngene™ CONVERTBLUE2126

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Blue Light Converter Screen converts UV to blue light for imaging of Safe Dyes. Use with UV transilluminators.

  • For use with Gbox Mini, NuGenius, Ingenius.

Specifications

Product Line Syngene
For Use With (Application) UV to blue light conversion
Color Blue
For Use With (Equipment) Gbox Mini, Nugenius, Ingenius
Quantity 1 Ea.
Size 21 x 26 cm
Voltage 100/115/220/230 V
Product Type Convertor Screen
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.