missing translation for 'onlineSavingsMsg'
Learn More

BLU9931, MedChemExpress

Product Code. 16488087
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16488087 5 mg 5mg
16475996 100 mg 100mg
16441567 50 mg 50mg
16405378 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16488087 Supplier Medchem Express Supplier No. HY12823

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress BLU9931 is a potent, highly selective, and irreversible fibroblast growth factor receptor 4 (FGFR4) inhibitor with an IC50 of 3 nM and a Kd of 6 nM. BLU9931 has significant antitumor activity.

BLU9931 is a potent, highly selective, and irreversible fibroblast growth factor receptor 4 (FGFR4) inhibitor with an IC50 of 3 nM and a Kd of 6 nM. BLU9931 has significant antitumor activity.

Specifications

Chemical Name or Material BLU9931
CAS 1538604-68-0
Color White
Purity Grade Notes Research
Molecular Formula C26H22Cl2N4O3
Quantity 5 mg
Solubility Information DMSO : 125 mg/mL (245.40 mM; Need ultrasonic)
SMILES C=CC(NC1=CC=CC(C)=C1NC2=NC=C3C=C(C4=C(Cl)C(OC)=CC(OC)=C4Cl)C=CC3=N2)=O
Molecular Weight (g/mol) 509.38
Formula Weight 509.38
Percent Purity 98.0%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.