missing translation for 'onlineSavingsMsg'
Learn More

BLU9931, MedChemExpress

Product Code. 16405378
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16405378

Brand: Medchem Express HY12823

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress BLU9931 is a potent, highly selective, and irreversible fibroblast growth factor receptor 4 (FGFR4) inhibitor with an IC50 of 3 nM and a Kd of 6 nM. BLU9931 has significant antitumor activity.

BLU9931 is a potent, highly selective, and irreversible fibroblast growth factor receptor 4 (FGFR4) inhibitor with an IC50 of 3 nM and a Kd of 6 nM. BLU9931 has significant antitumor activity.

Specifications

Chemical Name or Material BLU9931
CAS 1538604-68-0
Color White
Purity Grade Notes Research
Molecular Formula C26H22Cl2N4O3
Quantity 10 mg
Solubility Information DMSO : 125 mg/mL (245.40 mM; Need ultrasonic)
SMILES C=CC(NC1=CC=CC(C)=C1NC2=NC=C3C=C(C4=C(Cl)C(OC)=CC(OC)=C4Cl)C=CC3=N2)=O
Molecular Weight (g/mol) 509.38
Formula Weight 509.38
Percent Purity 98.0%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.