Learn More
Novus Biologicals™ Blu10 Plus Prestained Protein Ladder
Product Code. 30165442
Change view
Click to view available options
Quantity:
500 μL
Unit Size:
500µL
Product Code. Quantity unitSize
30165442 500 μL 500µL
1 options
Product Code. 30165442 Supplier Novus Biologicals™ Supplier No. NBP333170
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
Prestained Protein Ladder
The Blu10 Plus Prestained Protein Ladder is a combination of 10 pre-stained proteins with molecular weights from 6.5 to 270 kDa. The 10 recombinant proteins are covalently coupled with blue chromophore, while 2 orange bands at 30 kDa and 270 kDa and a green band at 52 kDa serve as reference bands. The Blu10 Plus Prestained Protein Ladder keeps track of the size and separation of proteins during SDS-polyacrylamide gel electrophoresis, approximating protein size and validating Western transfer efficiency on PVDF, nylon, or nitrocellulose membranes.
TRUSTED_SUSTAINABILITY
Specifications
Formulation 62.5 mM Tris-H3PO4 (pH 7.5), 1 mM EDTA, 2% SDS, 10 mM DTT, 33% glycerol
For Use With (Application) 3 μL or 5 μL per loading for clear visualization during electrophoresis on 15-well or 10-well mini-gel, respectively. 2.5 μL per well for general Western transferring.
Quantity 500 μL