missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ BLOTTO in TBS
Blocks non-specific binding site for antibodies on the membrane
Brand: Alfa Aesar™ J63238.AK
Code : NEW
Additional Details : Weight : 0.00025kg
Specifications
Blocks non-specific binding site for antibodies on the membrane | |
Blocking Buffer | |
−30° to −10°C |
Liquid | |
250mL |