missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies BloodStor™ 55-5

Product Code. 17169921
Encompass_Preferred
Change view
Click to view available options
Quantity:
16 x 7.2 mL
Unit Size:
Pack of 16
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17169921 16 x 7.2 mL Pack of 16
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17169921 Supplier STEMCELL Technologies Supplier No. 07937
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Optimized biopreservation reagent for hematopoietic cells and tissues

  • BloodStor™ is a series of generic cGMP freeze media products used to cryopreserve stem cells and other cells isolated from umbilical cord blood, peripheral blood, bone marrow, and other biologics
  • All BloodStor™ products are formulated using USP grade components and water for injection (WFI) quality water
  • BloodStor™ 55-5 is pre-formulated with 55% (w/v) DMSO USP, 5% (w/v) Dextran-40 USP, and WFI quality water

Specifications

Cell Type Hematopoietic Stem and Progenitor Cells
For Use With (Application) Cryopreservation
Quantity 16 x 7.2 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.