missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies BloodStor™ 100

Product Code. 17129931
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mL
100 mL
5 x 100 mL
Unit Size:
100mL
50mL
Pack of 5
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17129931 50 mL 50mL
17179921 5 x 100 mL Pack of 5
17189921 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17129931 Supplier STEMCELL Technologies Supplier No. 07951
 Restricted Item
Estimated Shipment: 23-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Biopreservation reagent for cells and tissues

  • BloodStor™ is a series of generic cGMP freezing media products used to cryopreserve stem cells and other cells isolated from umbilical cord blood, peripheral blood, bone marrow, and other biologics
  • BloodStor™ 100 contains 100% (w/v) DMSO USP

Specifications

Cell Type Hematopoietic Stem and Progenitor Cells
For Use With (Application) Cryopreservation
Quantity 50 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.