missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Blood group H inhibitor Rabbit anti-Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£359.00
Specifications
| Antigen | Blood group H inhibitor |
|---|---|
| Dilution | Western Blot 1.0 ug/ml |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
Blood group H inhibitor Polyclonal specifically detects Blood group H inhibitor in Mouse samples. It is validated for Western Blot.Specifications
| Blood group H inhibitor | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Mouse | |
| alpha (1,2) fucosyltransferase, Alpha(1,2)FT 1,2-alpha-L-fucosyltransferase, Blood group H alpha 2-fucosyltransferase, EC 2.4.1.69, Fucosyltransferase 1, fucosyltransferase 1 (galactoside 2-alpha-L-fucosyltransferase), fucosyltransferase 1 (galactoside 2-alpha-L-fucosyltransferase, Bombayphenotype included), fucosyltransferase 1 (galactoside 2-alpha-L-fucosyltransferase, H blood group), galactoside 2-alpha-L-fucosyltransferase 1, GDP-L-fucose:beta-D-galactoside 2-alpha-L-fucosyltransferase 1, HHH, HSC | |
| The immunogen is a synthetic peptide directed towards the C terminal region of mouse Blood group H inhibitor. Peptide sequence ALLTQCNHTIMTIGTFGFWAAYLAGGDTVYLANFTLPDSEFLKIFRPEAA | |
| Affinity purified |
| Western Blot 1.0 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS buffer, 2% sucrose | |
| 2523 | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title