missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Blood Group H Antigen Monoclonal Antibody (97-I)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA17675

Product Code. 11505372

  • £430.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

MA17675 is an anti-H INH blood antigen antibody that can be used to detect the the H H antigen of red blood cells and body fluids. It can be used for the testing of I-antigen in blood, blood stains, saliva, and for spot analysis in forensic medicine by absorption-elution, ELISA, and Inhibition assays.

The H antigen is a precursor to the ABO blood groupantigens, present in people of all common blood types. The Bombay phenotype (hh) does not express antigen H on red blood cells, and therefore this type will also lack A or B antigens, similar to the O blood group. However, unlike O group, the H antigen is absent, hence the individuals produce isoantibodies to antigen H as well as to both A and B antigens. If they receive blood from someone with O blood group, the anti-H antibodies will bind to the H antigen on the red blood cells of the donor blood and destroy the RBCs bycomplement-mediated lysis. Therefore, people with Bombay phenotype can receive blood only from otherhhdonors, although they can donate as though they were type O. Some individuals with the blood group A1 may also be able to produce anti-H antibodies due to the complete conversion of all the H antigen to A1 antigen. Production of the H antigen, or its deficiency in the Bombay phenotype, is controlled at the H locus on chromosome 19. The H locus contains threeexonsthat span more than 5kbof genomic DNA, and encodes thefucosyltransferasethat produces the H antigen on RBCs. The H antigen is a carbohydrate sequence withcarbohydrateslinked mainly to protein (with a minor fraction attached toceramidemoiety).
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Blood Group H Antigen
Monoclonal
1 mg/mL
PBS with 0.01% sodium azide; pH 7.2
Mouse
Protein A
RUO
Human
Antibody
IgM
Agglutination, ELISA, Inhibition Assays
97-I
Unconjugated
ABH; BG-H; BG-H1; group h
H human blood protein.
100 μg
Primary
4°C short term, -80°C long term
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.