missing translation for 'onlineSavingsMsg'
Learn More

Blood Group Antigen H (O) Type 1 Monoclonal Antibody (17-206), eBioscience™
GREENER_CHOICE

Product Code. 15298177
This item is not returnable. View return policy
Product Code. 15298177 Supplier Affymetrix eBioscience Supplier No. 14981082

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Blood Group Antigen H (O) Type 1 Monoclonal Antibody (17-206), eBioscience™

Blood Group Antigen H (O) Type 1 Monoclonal Antibody for IHC (P), ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen Blood Group Antigen H (O) Type 1
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 17-206
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene Alias ABH; BG-H; BG-H1; group h
Host Species Mouse
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.