missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Blood Group ABH Antigen Monoclonal Antibody (Z5H-2/Z2A)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA17667
This item is not returnable.
View return policy
Description
MA1-7667 detects AB human blood protein from human samples. MA1-7667 has been successfully used in agglutination applications. The MA1-7667 immunogen is AB human blood protein.
Blood-group antigens are generally defined as molecules formed by sequential addition of saccharides to the carbohydrate side chains of lipids and proteins detected on erythrocytes and certain epithelial cells. The A, B and H antigens are reported to undergo modulation during malignant cellular transformation. Blood group related antigens represent a group of carbohydrate determinants carried on both glycolipids and glycoproteins. They are usually mucin-type, and are detected on erythrocytes, certain epithelial cells, and in secretions of certain individuals. Sixteen genetically and biosynthetically distinct but inter-related specificities belong to this group of antigens, including A, B, H, Lewis A, Lewis B, Lewis X, Lewis Y, and precursor type 1 chain antigens.
Specifications
| Blood Group ABH Antigen | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1.25mM EDTA, 0.3M NaCl and 0.1% sodium azide; pH 7.2 | |
| Mouse | |
| Ammonium sulfate precipitation | |
| RUO | |
| Human | |
| Antibody | |
| IgM |
| Agglutination | |
| Z5H-2/Z2A | |
| Unconjugated | |
| ABH; BG-H; BG-H1 | |
| AB human blood protein. | |
| 100 μg | |
| Primary | |
| 4°C short term, -80°C long term | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction