missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Blood Group A Antigen Monoclonal Antibody (9A)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA191429
This item is not returnable.
View return policy
Description
MA1-91429 detects Blood Group A antigen from human samples.
Blood-group antigens are generally defined as molecules formed by sequential addition of saccharides to the carbohydrate side chains of lipids and proteins detected on erythrocytes and certain epithelial cells. The A, B and H antigens are reported to undergo modulation during malignant cellular transformation. Blood group related antigens represent a group of carbohydrate determinants carried on both glycolipids and glycoproteins. They are usually mucin-type, and are detected on erythrocytes, certain epithelial cells, and in secretions of certain individuals. Sixteen genetically and biosynthetically distinct but inter-related specificities belong to this group of antigens, including A, B, H, Lewis A, Lewis B, Lewis X, Lewis Y, and precursor type 1 chain antigens.
Specifica
| Blood Group A Antigen | |
| Monoclonal | |
| 1.0 mg/mL | |
| PBS with no preservative | |
| P16442 | |
| ABO | |
| Cell preparation: A431 cells (human vulval squamous carcinoma cell line). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG3 |
| ELISA, Immunoprecipitation | |
| 9A | |
| Unconjugated | |
| ABO | |
| A transferase; A1-specific alpha 1-3-N-acetylgalactosaminyltransferase; A3GALNT; A3GALT1; ABO; ABO A3 transferase; ABO blood group (transferase A, alpha 1-3-N-acetylgalactosaminyltransferase; transferase B, alpha 1-3-galactosyltransferase); ABO glycosyltransferase; ABO weak transfer; B transferase; B(A) alpha-1,3-galactosyltransferase; Blood Group A1; Blood Group A2; Blood Group A3; fucosylglycoprotein; fucosylglycoprotein 3-alpha-galactosyltransferase; Fucosylglycoprotein alpha-N-acetylgalactosaminyltransferase; Fucosylglycoprotein alpha-N-acetylgalactosaminyltransferase soluble form; Glycoprotein-fucosylgalactoside alpha-galactosyltransferase; glycoprotein-fucosylgalactoside alpha-N-acetylgalactosaminyltransferase; glycosyltransferase A; glycosyltransferase B; GTB; histo-blood group A transferase; histo-blood group A2 transferase; histo-blood group ABO system transferase; Histo-blood group B transferase; NAGAT; Transferase; transferase A | |
| Mouse | |
| IgG fraction | |
| RUO | |
| 28 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto