missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Blonanserin, MedChemExpress
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
25mg
Description
Blonanserin (AD-5423) is a potent and orally active 5-HT2A (Ki=0.812 nM) and dopamine D2 receptor (Ki =0.142 nM) antagonist. Blonanserin is usually acts as an atypical antipsychotic agent and can be used for the research of extrapyramidal symptoms, excessive sedation, or hypotension.
Specifications
Specifications
| Chemical Name or Material | Blonanserin |
| CAS | 132810-10-7 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C23H30FN3 |
| Quantity | 10 mg |
| Synonym | AD-5423 |
| Solubility Information | DMSO : 14.29 mg/mL (38.88 mM; Need ultrasonic) |
| SMILES | FC1=CC=C(C2=C(CCCCCC3)C3=NC(N4CCN(CC)CC4)=C2)C=C1 |
| Molecular Weight (g/mol) | 367.5 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction