missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Breaker Blocker Components - Blocking Bar Holder

Product Code. 11701186
Change view
Click to view available options
Color:
Yellow
Unit Size:
1 pack
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
11701186 Yellow 1 pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11701186 Supplier Brady™ Supplier No. 051265

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

For use with Bradys 480/600V Breaker Blocker

Specifications

Product Type Blocking Bar Holder
Alternate Part Number BREAKER BLOCKER ARM HOLDER 5 PACK
Color Yellow
Description Breaker Blocker Components - Blocking Bar Holder
Quantity 1 Ea.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.