missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BLOCK-iT™ Transfection Kit
Description
The BLOCK-iT™ Transfection Kit contains everything you need for successful high-efficiency transfection of siRNA into cells. Each kit contains Lipofectamine™ 2000 Transfection Reagent for efficient delivery in a broad range of cell lines and the BLOCK-iT™ Fluorescent Oligo for use as an indicator of transfection efficiency. The BLOCK-iT™ Fluorescent Oligo provides:
–A strong, easily detectable fluorescein signal indicating transfection efficiency
–Chemical modifications for a clearer, more persistent cellular signal than labeled siRNA
–Proven correlation of transfection efficiency with Stealth RNAi™ siRNA and traditional siRNA molecules
–A strong, easily detectable fluorescein signal indicating transfection efficiency
–Chemical modifications for a clearer, more persistent cellular signal than labeled siRNA
–Proven correlation of transfection efficiency with Stealth RNAi™ siRNA and traditional siRNA molecules
Specifications
Specifications
| Content And Storage | Each kit contains 125 μl of 20 μM BLOCK-iT™ Fluorescent Oligo and 250 μl of Lipofectamine™ 2000 Reagent. Store the BLOCK-iT™ Fluorescent Oligo at -20°C and Lipofectamine™ 2000 at +4°C |
| Control Type | Transfection Control |
| Format | Kit |
| Sample Type | Synthetic siRNA |
| Transfection Technique | Lipid-based Transfection |
| For Use With (Application) | Transfection |
| Product Line | BLOCK-iT |
| Product Type | Transfection Kit |
| Quantity | 1 Kit |
| Reagent Type | RNA |
For Research Use Only. Not for use in diagnostic procedures.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?