missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BLOC1S2 Monoclonal Antibody (03)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA530090
This item is not returnable.
View return policy
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human BLOC1S2/BLOS2.
BLOC1S2 is a component of the ubiquitously expressed BLOC1 multisubunit protein complex. BLOC1 is required for normal biogenesis of specialized organelles of the endosomal-lysosomal system, such as melanosomes and platelet dense granules.
Specifications
| BLOC1S2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| Q6QNY1 | |
| BLOC1S2 | |
| Recombinant Human BLOC1S2/BLOS2 protein (aa 1-99). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA | |
| 03 | |
| Unconjugated | |
| BLOC1S2 | |
| 11 kDa centrosome associated protein; 2410089B13Rik; biogenesis of lysosomal organelles complex 1 subunit 2; biogenesis of lysosomal organelles complex-1, subunit 2; biogenesis of lysosome-related organelles complex 1 subunit 2; biogenesis of lysosome-related organelles complex-1, subunit 2; biogenesis of lysosome-related organelles complex-1, subunit 2A; BLOC-1 subunit 2; BLOC1S2; Bloc1s2a; BLOS2; BORCS2; CEAP; CEAP11; centrosomal 10 kDa protein; centrosome protein oncogene; Centrosome-associated protein; RP11-316M21.4 | |
| Mouse | |
| Protein A | |
| RUO | |
| 282991 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction