missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BLOC1S2 Antibody (03), Alexa Fluor™ 700 , Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-06213AF700
This item is not returnable.
View return policy
Description
BLOC1S2 Monoclonal antibody specifically detects BLOC1S2 in Human samples. It is validated for ELISA
Specifications
| BLOC1S2 | |
| Monoclonal | |
| Alexa Fluor 700 | |
| biogenesis of lysosomal organelles complex-1, subunit 2, BLOC-1 subunit 2, CEAP, centrosomal 10 kDa protein, centrosome protein oncogene, Centrosome-associated protein, subunit 2 | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human BLOC1S2 (Q6QNY1-2; Met1-Arg99). | |
| 0.1 mL | |
| Cell Biology, Cell Cycle and Replication, Cytoskeleton Markers, Signal Transduction | |
| 282991 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| 03 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction