missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BLOC1S1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 19687-1-AP-20UL
This item is not returnable.
View return policy
Description
The antibody is specific to NLOC1S1.
BLOC1S1 is a component of the ubiquitously expressed BLOC1 multisubunit protein complex. BLOC1 is required for normal biogenesis of specialized organelles of the endosomal-lysosomal system, such as melanosomes and platelet dense granules (Starcevic and Dell′Angelica, 2004 ).Specifications
| BLOC1S1 | |
| Polyclonal | |
| Unconjugated | |
| BLOC1S1 | |
| BLOC 1 subunit 1, BLOC1S1, BLOS1, GCN5 like protein 1, GCN5L1, MICoA, Protein RT14, RT14 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 14533, 2647, 288785 | |
| -20°C | |
| Liquid |
| Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| 0.17 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| D3ZKU7, O55102, P78537 | |
| Bloc1s1 | |
| Peptide | |
| 20 μL | |
| Primary | |
| Mouse, Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction