missing translation for 'onlineSavingsMsg'
Learn More

BLOC1S1 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Product Code. 18395156 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18395156 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18395156 Supplier Novus Biologicals Supplier No. NBP304927100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

BLOC1S1 Polyclonal specifically detects BLOC1S1 in Human, Mouse samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BLOC1S1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias biogenesis of lysosomal organelles complex-1, subunit 1, BLOC subunit 1, BLOC-1 subunit 1, BLOS1MICoA, FLJ39337, FLJ97089, GCN5 (general control of amino-acid synthesis, yeast, homolog)-like 1, GCN5 general control of amino-acid synthesis 5-like 1, GCN5 general control of amino-acid synthesis 5-like 1 (yeast), GCN5L1biogenesis of lysosome-related organelles complex 1 subunit 1, GCN5-like protein 1, MGC87455, MTA1-interacting coactivator, Protein RT14, RT14
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 29-153 of human BLOC1S1 (NP_001478.2).
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2647
Target Species Human, Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.