missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ BLOC1S1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17180234 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17180234 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17180234 Supplier Invitrogen™ Supplier No. PA5112377

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

BLOC1S1 Polyclonal Antibody for ELISA, ICC/IF, IHC (P), Western Blot

BLOC1S1 is a component of the ubiquitously expressed BLOC1 multisubunit protein complex. BLOC1 is required for normal biogenesis of specialized organelles of the endosomal-lysosomal system, such as melanosomes and platelet dense granules (Starcevic and Dell'Angelica, 2004 ).
TRUSTED_SUSTAINABILITY

Specifications

Antigen BLOC1S1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.7 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene Bloc1s1
Gene Accession No. P78537
Gene Alias AI839753; biogenesis of lysosomal organelles complex 1 subunit 1; biogenesis of lysosomal organelles complex-1, subunit 1; biogenesis of lysosome-related organelles complex 1 subunit 1; biogenesis of lysosome-related organelles complex 1 subunit 1-like; biogenesis of lysosome-related organelles complex-1, subunit 1; BLOC subunit 1; BLOC-1 subunit 1; BLOC1S1; BLOS1; BORCS1; GCN5 (general control of amino-acid synthesis, yeast, homolog)-like 1; GCN5 general control of amino acid synthesis-like 1; GCN5 general control of amino-acid synthesis 5-like 1; Gcn5l1; GCN5-like protein 1; general control of amino acid synthesis, yeast homolog-like 1; general control of amino acid synthesis-like 1; MICoA; MTA1-interacting coactivator; Protein RT14; RT14; si:dkey-102c8.4; Unknown (protein for MGC:137062)
Gene Symbols Bloc1s1
Host Species Rabbit
Immunogen Recombinant Human Biogenesis of lysosome-related organelles complex 1 subunit 1 protein (1-153AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2647
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.