Learn More
BLNK (phospho Y84) Rabbit anti-Human, Mouse, Rat, Polyclonal , Abnova™
Rabbit polyclonal antibody raised against synthetic phosphopeptide of human BLNK.
Brand: Abnova PAB29646.100uL
Description
This gene encodes a cytoplasmic linker or adaptor protein that plays a critical role in B cell development. This protein bridges B cell receptor-associated kinase activation with downstream signaling pathways, thereby affecting various biological functions. The phosphorylation of five tyrosine residues is necessary for this protein to nucleate distinct signaling effectors following B cell receptor activation. Mutations in this gene cause hypoglobulinemia and absent B cells, a disease in which the pro- to pre-B-cell transition is developmentally blocked. Deficiency in this protein has also been shown in some cases of pre-B acute lymphoblastic leukemia. Alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Specifications
| BLNK (phospho Y84) | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic phosphopeptide of human BLNK. | |
| In PBS (without Mg2+ and Ca2+), 150 mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide) | |
| BLNK | |
| Synthetic phosphopeptide (conjugated with KLH) corresponding to residues surrounding Y84 of human BLNK. | |
| 100 μL | |
| Primary | |
| BLNK (phospho Y84) polyclonal antibody detects endogenous levels of human BLNK only when phosphorylated at tyrosine 84. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:100) Western Blot (1:500-1:3000) The optimal working dilution should be determined by the end user. | |
| BASH/BLNK-S/LY57/MGC111051/SLP-65/SLP65 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 29760 | |
| Human, Mouse, Rat | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.